(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%

Supplier: Ambeed
Danger

Synonyms: 2,2-Difluoro-1-methylcyclopropanecarboxylic acid

A436790-1G A436790-250MG A436790-5G A436790-100MG
77252-540EA 118 USD
77252-540 77252-542 77252-544 77746-890
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆F₂O₂
MDL Number: MFCD07777163
CAS Number: 128073-33-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR