3-Nitro-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Danger

A436025-1G A436025-250MG
77291-564EA 41.2 USD
77291-564 77291-566
3-Nitro-1H-pyrrolo[2,3-b]pyridine 98%
3-Nitro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD08272225
CAS Number: 23709-47-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR