2-Fluoroisobutyric acid 97%

Supplier: Ambeed
Danger

A435891-1G A435891-25G A435891-5G
77264-926EA 16.48 USD
77264-926 77264-928 77264-930
2-Fluoroisobutyric acid 97%
2-Fluoroisobutyric acid

Precaución: for Research Use Only

Formula: C₄H₇FO₂
MW: 106.1 g/mol
MDL Number: MFCD03452788
CAS Number: 63812-15-7
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR