3,6-Dibromo-2,7-dihydroxynaphthalene 97%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dibromonaphthalene-2,7-diol

A435677-1G A435677-250MG A435677-25G A435677-5G
77279-076EA 48.41 USD
77279-076 77279-078 77279-080 77279-082
3,6-Dibromo-2,7-dihydroxynaphthalene 97%
3,6-Dibromo-2,7-dihydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂O₂
MW: 317.98 g/mol
MDL Number: MFCD00277643
CAS Number: 96965-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR