2,6-Dichlorophenolindophenol sodium salt hydrate 95% (contains =< 11% H₂O)
Supplier: Ambeed
|
Warning
|
A435289-1G
A435289-25G
A435289-5G
77252-526EA
20
USD
77252-526
77252-528
77252-530
2,6-Dichlorophenolindophenol sodium salt hydrate 95% (contains =< 11% H₂O)
2,6-Dichlorophenolindophenol sodium salt hydrate
Precaución: for Research Use Only
Formula:
C₁₂H₆Cl₂NNaO₂·xH₂O |
MDL Number:
MFCD00150014 CAS Number: 1266615-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...