Ethyl 8-bromoisoquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

A435265-100MG A435265-1G A435265-250MG
77264-920EA 116 USD
77264-920 77264-922 77264-924
Ethyl 8-bromoisoquinoline-3-carboxylate 97%
Ethyl 8-bromoisoquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₂
MDL Number: MFCD28050435
CAS Number: 1823315-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR