5,10,15,20-tetrakis(p-chlorophenyl)porphyrin iron(III) chloride 97%

Supplier: Ambeed
Warning

A435114-100MG A435114-1G A435114-250MG A435114-5G
77252-520EA 21.63 USD
77252-520 77252-522 77252-524 77746-886
5,10,15,20-tetrakis(p-chlorophenyl)porphyrin iron(III) chloride 97%
5,10,15,20-tetrakis(p-chlorophenyl)porphyrin iron(III) chloride

Precaución: for Research Use Only

Formula: C₄₄H₂₄Cl₅FeN₄
MW: 841.79 g/mol
MDL Number: MFCD31537210
CAS Number: 36965-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR