5-((2S,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1-methylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

Synonyms: N1-Methylpseudouridine

A434807-100MG A434807-250MG A434807-50MG A434807-1G A434807-5G
77264-908EA 154.5 USD
77264-908 77264-910 77264-912 77551-302 77568-870
5-((2S,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1-methylpyrimidine-2,4(1H,3H)-dione 97%
5-((2S,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1-methylpyrimidine-2,4(1H,3H)-dione

Precaución: For research use only.

Formula: C₁₀H₁₄N₂O₆
MW: 258.22 g/mol
MDL Number: MFCD10687017
CAS Number: 13860-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR