BX-471 98%

Supplier: Ambeed
Warning

A434794-10MG A434794-1MG A434794-25MG A434794-5MG A434794-50MG A434794-250MG
77240-920EA 65 USD
77240-920 77240-922 77240-924 77240-926 77746-883 77746-882
BX-471 98%
BX-471

Precaución: for Research Use Only

Formula: C₂₁H₂₄ClFN₄O₃
MDL Number: MFCD09859709
CAS Number: 217645-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR