5-Hydroxyindole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A434465-250MG A434465-5G
77264-904EA 26 USD
77264-904 77264-906
5-Hydroxyindole-2-carboxylic acid 97%
5-Hydroxyindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD00005615
CAS Number: 21598-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR