5,6-Dihydrodibenzo[b,f]azocine 98%

Supplier: Ambeed
Warning

A434458-100MG A434458-1G A434458-250MG A434458-25G A434458-5G
77264-894EA 13.39 USD
77264-894 77264-896 77264-898 77264-900 77264-902
5,6-Dihydrodibenzo[b,f]azocine 98%
5,6-Dihydrodibenzo[b,f]azocine

Precaución: for Research Use Only

Formula: C₁₅H₁₃N
MW: 207.27 g/mol
MDL Number: MFCD28144233
CAS Number: 23194-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR