(S)-1-[(S)-2-(Di-tert-butylphosphino)ferrocenyl]ethylbis(2-methylphenyl)phosphine 98%

Supplier: Ambeed
Warning

A434328-100MG A434328-1G A434328-250MG A434328-25G A434328-5G
77252-496EA 20 USD
77252-496 77252-498 77252-500 77252-502 77252-504
(S)-1-[(S)-2-(Di-tert-butylphosphino)ferrocenyl]ethylbis(2-methylphenyl)phosphine 98%
(S)-1-[(S)-2-(Di-tert-butylphosphino)ferrocenyl]ethylbis(2-methylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₃₄H₄₄FeP₂
MW: 570.51 g/mol
MDL Number: MFCD08561173
CAS Number: 849924-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR