cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
cis-Dichlorotetrakis(dimethyl sulphoxide)ruthenium
A434326-100MG
A434326-1G
A434326-250MG
A434326-5G
77252-488EA
41.2
USD
77252-488
77252-490
77252-492
77252-494
cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II) 98%
cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II)
Precaución: for Research Use Only
Formula:
C₈H₂₄Cl₂O₄RuS₄ MW: 484.49 g/mol |
MDL Number:
MFCD00050955 CAS Number: 59091-96-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...