2',6'-Dimethoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,6-Dimethoxyphenyl)ethan-1-one, 1-(2,6-Dimethoxyphenyl)ethanone

A434225-100G A434225-10G A434225-1G A434225-25G A434225-5G
77252-478EA 623.15 USD
77252-478 77252-480 77252-482 77252-484 77252-486
2',6'-Dimethoxyacetophenone 98%
2',6'-Dimethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00008729
CAS Number: 2040-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR