LLY-507 ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-2'-[4-[2-(3-methyl-1H-indol-1-yl)ethyl]-1-piperazinyl]-N-[3-(1-pyrrolidinyl)propyl]-[1,1'-biphenyl]-3-carboxamide, 5-Cyano-2'-(4-(2-(3-methyl-1H-indol-1-yl)ethyl)piperazin-1-yl)-N-(3-(pyrrolidin-1-yl)propyl)-[1,1'-biphenyl]-3-carboxamide

A434113-10MG A434113-1MG A434113-25MG A434113-5MG
77240-912EA 57.68 USD
77240-912 77240-914 77240-916 77240-918
LLY-507 ≥98%
LLY-507

Precaución: for Research Use Only

Formula: C₃₆H₄₂N₆O
MW: 574.80 g/mol
MDL Number: MFCD28902312
CAS Number: 1793053-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR