SAR7334 99+%

Supplier: Ambeed
Warning

A433794-10MG A433794-1MG A433794-50MG A433794-5MG
77264-886EA 69 USD
77264-886 77264-888 77264-890 77264-892
SAR7334 99+%
SAR7334

Precaución: for Research Use Only

Formula: C₂₁H₂₂ClN₃O
MDL Number: MFCD27992059
CAS Number: 1333210-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR