6-((2-(Dimethylamino)ethyl)amino)-3-hydroxy-7H-indeno[2,1-c]quinolin-7-one dihydrochloride 98%

Supplier: Ambeed
Warning

A433632-10MG A433632-1MG A433632-50MG A433632-5MG
77240-896EA 45 USD
77240-896 77240-898 77240-900 77240-902
6-((2-(Dimethylamino)ethyl)amino)-3-hydroxy-7H-indeno[2,1-c]quinolin-7-one dihydrochloride 98%
6-((2-(Dimethylamino)ethyl)amino)-3-hydroxy-7H-indeno[2,1-c]quinolin-7-one dihydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₁Cl₂N₃O₂
MW: 406.30 g/mol
MDL Number: MFCD00943251
CAS Number: 174634-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR