(S)-Naringenin 95%

Supplier: Ambeed
Warning

Synonyms: (S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, (2S)-5,7,4'-trihydroxyflavanone, (-)-Naringenin

A433484-100G A433484-25G A433484-5G
77252-472EA 124 USD
77252-472 77252-474 77252-476
(S)-Naringenin 95%
(S)-Naringenin

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₅
MW: 272.25 g/mol
MDL Number: MFCD00870553
CAS Number: 480-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR