CCT-245737 97+%

Supplier: Ambeed
Warning

A433196-1MG A433196-25MG A433196-5MG A433196-10MG
77264-880EA 25 USD
77264-880 77264-882 77264-884 77616-870
CCT-245737 97+%
CCT-245737

Precaución: for Research Use Only

Formula: C₁₆H₁₆F₃N₇O
MDL Number: MFCD30489721
CAS Number: 1489389-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR