2-Bromo-1,3-difluoro-5-iodobenzene 98%

Supplier: Ambeed
Warning

A432691-100G A432691-10G A432691-1G A432691-25G A432691-500G A432691-5G
77291-538EA 261.62 USD
77291-538 77291-540 77291-542 77291-544 77291-546 77291-548
2-Bromo-1,3-difluoro-5-iodobenzene 98%
2-Bromo-1,3-difluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂I
MW: 318.89 g/mol
MDL Number: MFCD06797752
CAS Number: 155906-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR