(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-acetamidohexanoic acid 97%

Supplier: Ambeed
Warning

A432506-250MG A432506-1G
77240-888EA 17 USD
77240-888 77616-866
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-acetamidohexanoic acid 97%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-acetamidohexanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₅
MDL Number: MFCD15141996
CAS Number: 320410-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR