Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)(1,5-cyclooctadiene)ruthenium(II) 98%

Supplier: Ambeed
Warning

A432155-100MG A432155-250MG A432155-1G
77252-468EA 38 USD
77252-468 77252-470 77746-876
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)(1,5-cyclooctadiene)ruthenium(II) 98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)(1,5-cyclooctadiene)ruthenium(II)

Precaución: for Research Use Only

Formula: C₃₀H₅₀O₄Ru
MW: 575.8000 g/mol
MDL Number: MFCD01073791
CAS Number: 329735-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR