1,3-Difluoro-4-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Difluoroiodobenzene, 2,4-Difluoro-1-iodobenzene

A430957-100G A430957-25G A430957-500G A430957-5G
77291-522EA 46.35 USD
77291-522 77291-524 77291-526 77291-528
1,3-Difluoro-4-iodobenzene 98%
1,3-Difluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂I
MW: 239.99 g/mol
MDL Number: MFCD00040151
CAS Number: 2265-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR