2-Bromo-3-phenylpropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-phenylpropanoic acid

A430868-1G A430868-25G
77264-864EA 27.81 USD
77264-864 77264-866
2-Bromo-3-phenylpropionic acid 98%
2-Bromo-3-phenylpropionic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.08 g/mol
MDL Number: MFCD00461796
CAS Number: 16503-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR