4-Methyl-1,3-dioxane 98%

Supplier: Ambeed
Danger

A430404-25G A430404-5G
77291-518EA 72.1 USD
77291-518 77291-520
4-Methyl-1,3-dioxane 98%
4-Methyl-1,3-dioxane

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MW: 102.1317 g/mol
MDL Number: MFCD00006569
CAS Number: 1120-97-4
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR