2,4-Dihydroxy-3,6-dimethylbenzoic acid 95%

Supplier: Ambeed
Warning

A430298-100MG A430298-250MG
77252-448EA 132 USD
77252-448 77252-450
2,4-Dihydroxy-3,6-dimethylbenzoic acid 95%
2,4-Dihydroxy-3,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD00239340
CAS Number: 4707-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR