BI-9564 98%

Supplier: Ambeed
Danger

A429986-10MG A429986-1MG A429986-25MG A429986-50MG A429986-5MG
77264-838EA 106 USD
77264-838 77264-840 77264-842 77264-844 77264-846
BI-9564 98%
BI-9564

Precaución: for Research Use Only

Formula: C₂₀H₂₃N₃O₃
MDL Number: MFCD30182324
CAS Number: 1883429-22-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR