Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate 99% Au 39.91 ± 0.8%

Supplier: Ambeed
Warning

A429915-100MG A429915-1G A429915-250MG A429915-5G
77252-436EA 103 USD
77252-436 77252-438 77252-440 77252-442
Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate 99% Au 39.91 ± 0.8%
Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₅₄H₄₈Au₃B₄F₁₆OP₃
MW: 1743.9977 g/mol
MDL Number: MFCD06658141
CAS Number: 53317-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR