7-Methoxy-1H-benzo[d]imidazole 98%

Supplier: Ambeed
Warning

Synonyms: 7-Methoxy-1H-benzimidazole

A429834-250MG A429834-1G
77240-868EA 29.87 USD
77240-868 77616-848
7-Methoxy-1H-benzo[d]imidazole 98%
7-Methoxy-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD11973699
CAS Number: 27080-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR