Choline hydrogen tartrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate
A429761-100G
A429761-25G
A429761-500G
A429761-1000G
77252-430EA
27
USD
77252-430
77252-432
77252-434
77746-868
Choline hydrogen tartrate 98%
Choline hydrogen tartrate
Precaución: for Research Use Only
Formula:
C₉H₁₉NO₇ MW: 253.25 g/mol |
MDL Number:
MFCD00036332 CAS Number: 87-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...