4-Acetoxybenzoic acid 98%

Supplier: Ambeed
Warning

A429729-100G A429729-25G A429729-500G A429729-5G
77291-510EA 88.58 USD
77291-510 77291-512 77291-514 77291-516
4-Acetoxybenzoic acid 98%
4-Acetoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00002540
CAS Number: 2345-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR