4-Cyanobenzoyl chloride 98%

Supplier: Ambeed
Danger

A429652-10G A429652-1G A429652-5G A429652-25G
77291-504EA 39 USD
77291-504 77291-506 77291-508 77594-334
4-Cyanobenzoyl chloride 98%
4-Cyanobenzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₄ClNO
MDL Number: MFCD00001822
CAS Number: 6068-72-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR