2-Cyano-4'-fluoroacetanilide 95%

Supplier: Ambeed
Warning

A429407-1G A429407-250MG A429407-5G
77264-832EA 15 USD
77264-832 77264-834 77264-836
2-Cyano-4'-fluoroacetanilide 95%
2-Cyano-4'-fluoroacetanilide

Precaución: for Research Use Only

Formula: C₉H₇FN₂O
MDL Number: MFCD00276110
CAS Number: 1735-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR