4-(3-(Hydroxymethyl)phenoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A429026-100MG A429026-1G A429026-250MG
77264-826EA 154 USD
77264-826 77264-828 77264-830
4-(3-(Hydroxymethyl)phenoxy)benzonitrile 97%
4-(3-(Hydroxymethyl)phenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₂
MDL Number: MFCD16500081
CAS Number: 888967-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR