4-(Trifluoromethylthio)benzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: (4-(Bromomethyl)phenyl)(trifluoromethyl)sulphane

A428853-250MG A428853-5G
77291-486EA 18 USD
77291-486 77291-488
4-(Trifluoromethylthio)benzyl bromide 97%
4-(Trifluoromethylthio)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₆BrF₃S
MDL Number: MFCD00129189
CAS Number: 21101-63-3
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR