MLN-9708 99%

Supplier: Ambeed
Warning

Synonyms: Ixazomib, MLN9708, 4-(Carboxymethyl)-2-((R)-1-(2-(2,5-dichlorobenzamido)acetamido)-3-methylbutyl)-6-oxo-1,3,2-dioxaborinane-4-carboxylic acid

A428391-100MG A428391-10MG A428391-1MG A428391-5MG A428391-50MG
77240-860EA 116 USD
77240-860 77240-862 77240-864 77240-866 77746-867
MLN-9708 99%
MLN-9708

Precaución: for Research Use Only

Formula: C₂₀H₂₃BCl₂N₂O₉
MDL Number: MFCD18251437
CAS Number: 1201902-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR