Diethylene Glycol bis(2-propynyl) ether 95%

Supplier: Ambeed
Danger

Synonyms: 3-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]-1-propyne, Bis-propargyl-PEG3, 4,7,10-Trioxa-1,12-tridecadiyne, 3-(2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethoxy)prop-1-yne

A427760-100MG A427760-1G A427760-250MG A427760-5G
77264-816EA 32.96 USD
77264-816 77264-818 77264-820 77264-822
Diethylene Glycol bis(2-propynyl) ether 95%
Diethylene Glycol bis(2-propynyl) ether

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
MDL Number: MFCD22201523
CAS Number: 126422-57-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR