Hydroxy-PEG3-CH2CO2tBu 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(2-(2-(2-hydroxyethoxy)ethoxy)ethoxy)acetate, Hydroxy-PEG3-CH2-Boc

A427638-100MG A427638-1G A427638-250MG
77264-812EA 51.5 USD
77264-812 77264-814 77748-864
Hydroxy-PEG3-CH2CO2tBu 98%
Hydroxy-PEG3-CH2CO2tBu

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₆
MW: 264.315 g/mol
MDL Number: MFCD27977510
CAS Number: 518044-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR