2,2-Dimethylcyclopropanecarboxylic acid ≥97%

Supplier: Ambeed
Danger

A427430-10G A427430-25G
77252-420EA 384.19 USD
77252-420 77252-422
2,2-Dimethylcyclopropanecarboxylic acid ≥97%
2,2-Dimethylcyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD04972493
CAS Number: 75885-59-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR