Tris[3-(trimethoxysilyl)propyl] Isocyanurate 98%

Supplier: Ambeed
Danger

Synonyms: 1,3,5-Tris(3-(trimethoxysilyl)propyl)-1,3,5-triazinane-2,4,6-trione, Isocyanuric acid tris[3-(trimethoxysilyl)propyl] ester

A426355-1G A426355-25G A426355-5G
77252-406EA 5.15 USD
77252-406 77252-408 77252-410
Tris[3-(trimethoxysilyl)propyl] Isocyanurate 98%
Tris[3-(trimethoxysilyl)propyl] Isocyanurate

Precaución: for Research Use Only

Formula: C₂₁H₄₅N₃O₁₂Si₃
MW: 615.86 g/mol
MDL Number: MFCD00054746
CAS Number: 26115-70-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR