((3AS,5R,6S,6aS)-6-hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)(morpholino)methanone 98%

Supplier: Ambeed
Warning

A426318-100MG A426318-1G A426318-250MG A426318-25G A426318-5G
77252-396EA 5.15 USD
77252-396 77252-398 77252-400 77252-402 77252-404
((3AS,5R,6S,6aS)-6-hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)(morpholino)methanone 98%
((3AS,5R,6S,6aS)-6-hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₆
MW: 273.28 g/mol
MDL Number: MFCD25977365
CAS Number: 1103738-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR