4,6-Dichloro-5-fluoronicotinic acid 98%

Supplier: Ambeed
Danger

A425921-100MG A425921-1G A425921-250MG A425921-5G
77252-388EA 11.33 USD
77252-388 77252-390 77252-392 77252-394
4,6-Dichloro-5-fluoronicotinic acid 98%
4,6-Dichloro-5-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FNO₂
MW: 209.99 g/mol
MDL Number: MFCD18256593
CAS Number: 154012-18-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR