2-(tert-Butylamino)ethanol 98%

Supplier: Ambeed
Warning

A425462-100G A425462-25G A425462-500G
77252-380EA 76 USD
77252-380 77252-382 77594-314
2-(tert-Butylamino)ethanol 98%
2-(tert-Butylamino)ethanol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MDL Number: MFCD00002837
CAS Number: 4620-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR