2-Iodo-3-nitrotoluene 97%

Supplier: Ambeed
Warning

A425395-100G A425395-10G A425395-1G A425395-25G A425395-5G
77264-774EA 228 USD
77264-774 77264-776 77264-778 77264-780 77264-782
2-Iodo-3-nitrotoluene 97%
2-Iodo-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆INO₂
MDL Number: MFCD00091040
CAS Number: 6277-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR