Methyl 4-((4-chlorobenzyl)oxy)thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A424970-1G A424970-250MG
77240-848EA 210.12 USD
77240-848 77240-850
Methyl 4-((4-chlorobenzyl)oxy)thiophene-2-carboxylate 97%
Methyl 4-((4-chlorobenzyl)oxy)thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO₃S
MW: 282.74 g/mol
MDL Number: MFCD26129653
CAS Number: 1708251-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR