4-Bromo-2-nitro(trifluoromethoxy)benzene 97%

Supplier: Ambeed
Warning

A424835-100G A424835-10G A424835-1G A424835-5G A424835-25G
77291-464EA 63 USD
77291-464 77291-466 77291-468 77291-470 77748-855
4-Bromo-2-nitro(trifluoromethoxy)benzene 97%
4-Bromo-2-nitro(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₃NO₃
MDL Number: MFCD04039243
CAS Number: 95668-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR