4,4'-Dimethoxybenzhydrol 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dimethoxy-α-phenylbenzyl alcohol, Bis(4-methoxyphenyl)methanol, Bis(4-methoxyphenyl)carbinol, 4,4'-Dimethoxyphenyl phenyl carbinol, 4,4'-Dimethoxybenzhydryl alcohol, 4,4'-Dimethoxydiphenylmethanol

A424522-1G A424522-25G A424522-5G
77291-448EA 11.33 USD
77291-448 77291-450 77291-452
4,4'-Dimethoxybenzhydrol 98%
4,4'-Dimethoxybenzhydrol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₃
MW: 244.29 g/mol
MDL Number: MFCD00008410
CAS Number: 728-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR