5-Fluoro-2,1,3-benzothiadiazole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluorobenzo[c][1,2,5]thiadiazole

A423839-1G A423839-250MG
77291-438EA 26 USD
77291-438 77291-440
5-Fluoro-2,1,3-benzothiadiazole 95%
5-Fluoro-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₆H₃FN₂S
MW: 154.1700 g/mol
MDL Number: MFCD00173946
CAS Number: 17821-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR