3,4-Dichloro-5-nitropyridine 98%

Supplier: Ambeed
Warning

A423453-1G A423453-25G A423453-5G
77278-988EA 97.85 USD
77278-988 77278-990 77278-992
3,4-Dichloro-5-nitropyridine 98%
3,4-Dichloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MW: 192.988 g/mol
MDL Number: MFCD09910258
CAS Number: 56809-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR