3-Chloro-6-piperidin-1-ylpyridazine 95%

Supplier: Ambeed
Warning

A423413-1G A423413-250MG
77278-984EA 18 USD
77278-984 77278-986
3-Chloro-6-piperidin-1-ylpyridazine 95%
3-Chloro-6-piperidin-1-ylpyridazine

Precaución: for Research Use Only

Formula: C₉H₁₂ClN₃
MDL Number: MFCD00574572
CAS Number: 1722-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR